missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £437.00
Specifications
| Antigen | Luciferase |
|---|---|
| Clone | 965808 |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710737
![]() |
R&D Systems
MAB10026-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753127
![]() |
R&D Systems
MAB10026-100 |
100 μg |
£437.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Luciferase Monoclonal specifically detects Luciferase in Multi-Species samples. It is validated for Western Blot.Specifications
| Luciferase | |
| Western Blot 2 μg/mL | |
| Monoclonal | |
| Lyophilized | |
| Multi-species | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 965808 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Synthetic peptide of NanoLuc™ (Nluc) Luciferase | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. Detects NanoLuc™ (Nluc) Luciferase and Large BiT in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title