missing translation for 'onlineSavingsMsg'
Learn More

NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18753127
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Product Code. Quantity unitSize
18753127 100 μg 100µg
18710737 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 18753127

Brand: R&D Systems MAB10026100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Luciferase Monoclonal specifically detects Luciferase in Multi-Species samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luciferase
Applications Western Blot
Classification Monoclonal
Clone 965808
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Mouse
Immunogen Synthetic peptide of NanoLuc™ (Nluc) Luciferase
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. Detects NanoLuc™ (Nluc) Luciferase and Large BiT in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.