missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (CL5810), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £419.00
Specifications
| Antigen | Nanog |
|---|---|
| Clone | CL5810 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285284
|
Novus Biologicals
NBP2-61429 |
100 μL |
£419.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668967
|
Novus Biologicals
NBP2-61429-25ul |
25 μL |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Nanog | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| NANOG | |
| IgG1 | |
| Protein A purified |
| CL5810 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Autophagy, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Markers, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway | |
| 79923 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: DSPDSSTSPKGKQPTSAEK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title