missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (CL5810), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61429
This item is not returnable.
View return policy
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nanog | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NANOG | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: DSPDSSTSPKGKQPTSAEK | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5810 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Mouse | |
| Protein A purified | |
| Apoptosis, Autophagy, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Markers, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway | |
| 79923 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction