missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (5A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
£178.00 - £400.00
Specifications
| Antigen | Nanog |
|---|---|
| Clone | 5A10 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283981
|
Novus Biologicals
NBP1-04320SS |
0.025 mL |
£178.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18747073
|
Novus Biologicals
NBP1-04320 |
0.1 mL |
£400.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Nanog | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| Q9H9S0 | |
| 79923 | |
| Recombinant human Nanog (1-154aa) purified from E. coli (NP_079141). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 35 kDa |
| 5A10 | |
| Western Blot 1:500-1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| NANOG | |
| IgG2a κ | |
| Protein G purified | |
| Anti-human Nanog mAb, clone 5A10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Nanog protein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title