missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (5A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-04320SS
This item is not returnable.
View return policy
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Nanog | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Q9H9S0 | |
| NANOG | |
| Recombinant human Nanog (1-154aa) purified from E. coli (NP_079141). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Nanog mAb, clone 5A10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Nanog protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5A10 | |
| Unconjugated | |
| PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Mouse | |
| 35 kDa | |
| 0.025 mL | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| 79923 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction