missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nanaocolor™ Nitrite 2 Tube Test

Product Code. 10179111 Shop All Macherey-Nagel Products
Change view
Click to view available options
No. of Tests:
2
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10179111 2 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10179111 Supplier Macherey-Nagel™ Supplier No. 985068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guarantees maximum safety for the user because measurement is performed directly in the test tube. Macherey Nagel™ Nanocolor™ Nitrite 2 Tube Tests automatically perform the measurement after the cuvette is inserted into the cuvette slot.

  • Photometric determination with sulphanilamide and N-(1-naphthyl)-ethylene diamine
  • Nitrite reacts with sulphanilamide and N-(1-naphthyl)-ethylene diamine (lyophilized) to a red-violet azo dye
  • Reaction basis based on international approved standard methods like DIN-, EN-, ISO-, EPA- and APHA
  • Tests contain accurate predosed reagents that are directly ready to use
  • Every test box is equipped with a color-coded label and can be identified clearly

Specifications

For Use With (Application) Photometric Determination of Nitrite
No. of Tests 2
compliance-icons
Product Identifier
  • NANOCOLOR Nitrite 2
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: sulfanilamide, citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.