missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nanaocolor™ Ammonium 3 Tube Tests

Product Code. 10772701 Shop All Macherey-Nagel Products
Change view
Click to view available options
Range:
0.04 to 2.30 mg/L NH4-N, 0.05 to 3.00 mg/L NH4/NH3
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10772701 0.04 to 2.30 mg/L NH4-N, 0.05 to 3.00 mg/L NH4/NH3 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10772701 Supplier Macherey-Nagel™ Supplier No. 985003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guarantees maximum safety for the user because measurement is performed directly in the test tube. Macherey Nagel™ Nanocolor™ Ammonium 3 Tube Tests automatically perform the measurement after the cuvette is inserted into the cuvette slot.

  • Photometric determination of phosphate
  • Reaction basis based on international approved standard methods like DIN-, EN-, ISO-, EPA- and APHA
  • Tests contain accurate predosed reagents that are directly ready to use
  • Every test box is equipped with a color-coded label and can be identified clearly

Specifications

Includes 20 test tubes Ammonium 3
1 tube Nanofix™ Ammonium 3 R2
1 test tube with blank value “NULL”
No. of Tests 20
Range 0.04 to 2.30 mg/L NH4-N, 0.05 to 3.00 mg/L NH4/NH3
Test Time 15 min.
compliance-icons compliance-icons
Product Identifier
  • NANOCOLOUR ammonium 3
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: sodium nitroprusside, dichloroisocyanuric acid, Na salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.