missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | NALP12 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655032
|
Novus Biologicals
NBP2-94058-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654942
|
Novus Biologicals
NBP2-94058-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NALP12 Polyclonal antibody specifically detects NALP12 in Human samples. It is validated for Western BlotSpecifications
| NALP12 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 91662 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 862-1061 of human NALP12 (NP_653288.1). WLKICRLTAAACDELASTLSVNQSLRELDLSLNELGDLGVLLLCEGLRHPTCKLQTLRLGICRLGSAACEGLSVVLQANHNLRELDLSFNDLGDWGLWLLAEGLQHPACRLQKLWLDSCGLTAKACENLYFTLGINQTLTDLYLTNNALGDTGVRLLCKRLSHPGCKLRVLWLFGMDLNKMTHSRLAALRVTKPYLDIGC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title