missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94058-0.02ml
This item is not returnable.
View return policy
Description
NALP12 Polyclonal antibody specifically detects NALP12 in Human samples. It is validated for Western Blot
Specifications
| NALP12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 862-1061 of human NALP12 (NP_653288.1). WLKICRLTAAACDELASTLSVNQSLRELDLSLNELGDLGVLLLCEGLRHPTCKLQTLRLGICRLGSAACEGLSVVLQANHNLRELDLSFNDLGDWGLWLLAEGLQHPACRLQKLWLDSCGLTAKACENLYFTLGINQTLTDLYLTNNALGDTGVRLLCKRLSHPGCKLRVLWLFGMDLNKMTHSRLAALRVTKPYLDIGC | |
| 0.02 mL | |
| Apoptosis | |
| 91662 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction