Learn More
Naloxone hydrochloride, 98%, Thermo Scientific™
Naloxone hydrochloride, CAS # 357-08-4, is involved in the blockade of the mu-opioid receptor and removal of other compounds from the receptor site.
£66.09 - £444.00
Chemical Identifiers
| CAS | 357-08-4 |
|---|---|
| Molecular Formula | C19H22ClNO4 |
| Molecular Weight (g/mol) | 363.838 |
| MDL Number | MFCD00069322 |
| InChI Key | RGPDIGOSVORSAK-STHHAXOLSA-N |
| Synonym | naloxone hydrochloride, naloxone hcl, narcan, evzio, unii-f850569pqr, nalonee, 17-allyl-4,5alpha-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride, narcan tn, naloxonehydrochloride |
| PubChem CID | 5464092 |
| IUPAC Name | (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride |
| SMILES | C=CCN1CCC23C4C(=O)CCC2(C1CC5=C3C(=C(C=C5)O)O4)O.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15452859
|
Thermo Scientific Alfa Aesar
J60013.MC |
100mg |
£67.60 £66.09 / 100mg Save £1.51 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15462859
|
Thermo Scientific Alfa Aesar
J60013.ME |
500mg |
£232.00 £221.01 / 500mg Save £10.99 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15442859
|
Thermo Scientific Alfa Aesar
J60013.03 |
1g |
£466.00 £444.00 / 1g Save £22.00 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 357-08-4 | |
| 363.838 | |
| RGPDIGOSVORSAK-STHHAXOLSA-N | |
| 5464092 | |
| C=CCN1CCC23C4C(=O)CCC2(C1CC5=C3C(=C(C=C5)O)O4)O.Cl |
| C19H22ClNO4 | |
| MFCD00069322 | |
| naloxone hydrochloride, naloxone hcl, narcan, evzio, unii-f850569pqr, nalonee, 17-allyl-4,5alpha-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride, narcan tn, naloxonehydrochloride | |
| (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride |
Specifications
| 357-08-4 | |
| MFCD00069322 | |
| naloxone hydrochloride, naloxone hcl, narcan, evzio, unii-f850569pqr, nalonee, 17-allyl-4,5alpha-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride, narcan tn, naloxonehydrochloride | |
| RGPDIGOSVORSAK-STHHAXOLSA-N | |
| (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride | |
| 5464092 | |
| 98% |
| C19H22ClNO4 | |
| 14,6362 | |
| Soluble in water (73mg/ml),ethanol (3.3mg/ml),methanol (50mg/ml) and dimethyl sulfoxide (73mg/ml). Insoluble in ether | |
| C=CCN1CCC23C4C(=O)CCC2(C1CC5=C3C(=C(C=C5)O)O4)O.Cl | |
| 363.838 | |
| 363.84 | |
| Naloxone hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 206-611-0
missing translation for 'rtecsNumber' : QD2275000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.