missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PFA Graduated Plastic Beakers with Handles
GREENER_CHOICE

Product Code. 10565781
Click to view available options
Capacity (Metric):
1000 mL
3000 mL
Unit Size:
Each
This item is not returnable. View return policy
Not for use on hotplates.

Product Code. 10565781

Brand: Thermo Scientific™ DS15113000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Not for use on hotplates.

Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with these PFA graduated beakers with handles.

Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handles. These beakers feature extraordinary chemical resistance. PFA is non-stick, making the beaker easy to clean and resistant to contamination.

  • Withstands exposure to extreme temperatures from -270° to +250°C
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded-in graduation in both oz. and mL won't wear off with continuous use or washings
  • Excellent chemical resistance to strong ozidizing agents, acids, hydrocarbons and ketones make these beakers useful for a broad range of high-purity applications
  • Molded-in rectangular handle for easy, safe handling and pouring
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Color Natural translucent
Description Beaker with Handle PFA 3000 mL
Capacity (Metric) 3000 mL
Material PFA
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Molded Dual Scale
Style Handled
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.