missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAIP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£141.00
Specifications
| Antigen | NAIP |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunoprecipitation, Immunomicroscopy, SDS-Page, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy |
| Classification | Polyclonal |
Description
NAIP Polyclonal specifically detects NAIP in Human, E. coli samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunomicroscopy, SDS-Page, Immunofluorescence, Immunocytochemistry.Specifications
| NAIP | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunoprecipitation, Immunomicroscopy, SDS-Page, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer | |
| PBS with 0.02% Sodium Azide | |
| baculoviral IAP repeat-containing 1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP | |
| NAIP | |
| IgG | |
| Affinity Purified | |
| Minor, lower molecular weight bands may represent alternately spliced forms. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy | |
| Unconjugated | |
| RUO | |
| Human, E. coli | |
| AAC52047 | |
| 4671 | |
| Antibody was raised against a synthetic peptide corresponding to 13 amino acids at the C-terminus of human NAIP. The immunogen is located within amino acids 1200 - 1250 of NAIP. Amino Acid Squence: QFPDEETSEKFAY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 155 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title