missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAIP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-77196-0.025mg
This item is not returnable.
View return policy
Description
NAIP Polyclonal specifically detects NAIP in Human, E. coli samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunomicroscopy, SDS-Page, Immunofluorescence, Immunocytochemistry.
Specifications
| NAIP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| baculoviral IAP repeat-containing 1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP | |
| Rabbit | |
| 155 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer | |
| 4671 | |
| Human, E. coli | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunoprecipitation, Immunomicroscopy, SDS-Page, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
| AAC52047 | |
| NAIP | |
| Antibody was raised against a synthetic peptide corresponding to 13 amino acids at the C-terminus of human NAIP. The immunogen is located within amino acids 1200 - 1250 of NAIP. Amino Acid Squence: QFPDEETSEKFAY | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Minor, lower molecular weight bands may represent alternately spliced forms. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction