missing translation for 'onlineSavingsMsg'
Learn More

Nafion™ R-1100 resin, sulfonyl fluoride form, Thermo Scientific™

Product Code. 11405440
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405440 250 g 250g
11475430 2 g 2g
11485430 10 g 10g
11495430 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11405440 Supplier Thermo Scientific Alfa Aesar Supplier No. 044626.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermoplastic-can be extruded or laminated

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26654-97-7
Molecular Formula C9F18O4S
Molecular Weight (g/mol) 546.126
MDL Number MFCD05665455
InChI Key DHYYUWYUPUOPFL-UHFFFAOYSA-N
Synonym 1,1,2,2-tetrafluoroethene; 1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-1,2,2-trifluoroethenoxy propan-2-yl oxyethanesulfonyl fluoride, 1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-trifluoroethenyl oxy propan-2-yl oxy ethanesulfonyl fluoride-tetrafluoroethene 1:1, tetrafluoroethylene-perfluoro sulfinoethoxypropoxytrifluoroethylene copolymer, ethanesulfonyl fluoride, 2-1-difluoro 1,2,2-trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer with 1,1,2,2-tetrafluoroethene, ethanesulfonyl fluoride, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer with tetrafluoroethene, nafion r r-1100 resin, sulfonyl fluoride form 1g, nafion r-1100 membrane, 0.05mm thick, sulfonyl fluoride form, 1,1,2,2-tetrakis fluoranyl ethene; 1,1,2,2-tetrakis fluoranyl-2-1,1,1,2,3,3-hexakis fluoranyl-3-1,2,2-tris fluoranyl ethenoxy propan-2-yl oxy-ethanesulfonyl fluoride
PubChem CID 117323
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonyl fluoride
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)F)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Specifications

Melting Point >200°C
Quantity 250 g
Solubility Information Insoluble in water.
Physical Form Pellets
Chemical Name or Material Nafion™ R-1100 resin, Sulfonyl fluoride form
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.