accessibility menu, dialog, popup

UserName

CAS RN 26654-97-7

FFFFFFFOFFFOFFFFSFOOFFF

CAS RN 26654-97-7

IUPAC Name: 1,1,2,2-tetrafluoro-2-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)ethane-1-sulfonyl fluoride; tetrafluoroethene
Synonyms: 1,1,2,2-tetrafluoro-2-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)ethanesulfonyl fluoride; tetrafluoroethylene
Molecular Weight (g/mol): 546.13
Molecular Formula: C9F18O4S
InChi Key: DHYYUWYUPUOPFL-UHFFFAOYNA-N
SMILES: FC(F)=C(F)F.FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)S(F)(=O)=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)