missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set

Product Code. 12694255
Click to view available options
Quantity:
1 set
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12694255

Brand: Invitrogen™ 4374348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Substrate, required buffer and reaction accelerator

Designed for rapid and sensitive quantitation of influenza neuraminidase inhibitor resistance in dilutions of virus culture medium in a 96-well microplate format. Thermo Scientific NA-Star Influenza Neuraminidase Inhibitor Reagent kit provides all necessa

NA-Star Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set includes everything needed to quantitate neuraminidase activity and neuraminidase inhibitor resistance in avian, equine, human (types A and B), and porcine influenza viruses. The kit's fast and easy protocol and convenient 96-well plate format make it ideal for monitoring influenza virus neuraminidase inhibitor resistance, as well as high-throughput inhibitor compound screening.

  • Includes pre-made solutions for all required reagents, enabling you to standardize assays and compare results between labs easily
  • Provides up to 50-fold higher sensitivity than assays using fluorescent methylumbelliferone N-acetylneuraminic acid (MUNANA) substrate
  • Dynamic range of four orders of magnitude enables you to quantitate virus isolates over a broad range of virus concentration and with varying levels of neuraminidase activity without performing numerous sample dilutions
  • Fast and easy protocol enables you to complete assays in less than 1.5 hours, providing rapid assay throughput

Specifications

  • Target: Neuraminidase
  • Detection Method: Enhanced Chemiluminescence
  • For use with Equipment: Luminometer (Microplate)
  • Form: Liquid
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Neuraminidase Substrate
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The set includes the substrate, required buffer, and reaction accelerator. Store all components at 2°C to 8°C.
Detection Method Enhanced Chemiluminescence
For Use With (Application) Influenza Neuraminidase Assay, Inhibitor Resistance Detection
For Use With (Equipment) Luminometer (Microplate)
Product Type Neuraminidase Assay
Form Liquid
Format 96-well plate
Product Line NA-Star, NovaBright
Quantity 1 set
Shipping Condition Wet Ice
Substrate NA-Star™
Substrate Properties Chemical Substrate
Substrate Type Neuraminidase Substrate
Target Enzyme Sialidase, Neuraminidase
Throughput High-throughput Compatible
Validated Application Enzyme Assay
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.