missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NA-Fluor™ Influenza Neuraminidase Assay Kit

Product Code. 15849724
Change view
Click to view available options
Quantity:
960 assays
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849724 960 assays Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15849724 Supplier Invitrogen™ Supplier No. 4457091
  • ONLINE EXCLUSIVE PRICE
    £806.00
    £709.65 / Pack of 960
    Save £96.35
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NA-Fluor MUNANA Substrate (4-(methylumbelliferyl)-N-acetylneuraminic acid), NA-Fluor 2X Assay Buffer and NA-Fluor Stop Solution

An assay for monitoring neuraminidase enzyme activity from mixed viral populations, non-viral or bacterial sources. Thermo Scientific NA-Fluor Influenza Neuraminidase Assay Kit provides validated reagents and standardized protocol for conducting neuramini

  • Efficient design - neuraminidase assay in one complete kit
  • Optimized Formulations - assay reagents have been optimized for maximum performance based on NISN protocols
  • Robust Application - functional detection of NI-resistant virus in mixed viral populations
  • Easy of use and Flexible Screening - stable fluorescent signal enables batch-mode or high throughput processing

Specifications

  • Color: Blue
  • Detection Method: Fluorescent
  • Filter Type: Emission 440-460, Excitation 350-365
  • For Use With Equipment: Fluorometer
  • Form: Lyophilized
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible

Neuraminidase Assay in One Complete Kit
The NA-Fluor Influenza Neuraminidase Assay Kit includes comprehensive protocols for titering viral isolates based upon neuraminidase activity and conducting neuraminidase enzyme inhibition assays. The assay can also be used for monitoring neuraminidase enzyme activity from non-viral or bacterial sources. The NA-Fluor Kit contains reagents for ten 96-well microplates sufficient for a total of 960 assay wells and includes the following components: fluorescent MUNANA neuraminidase substrate, assay buffer, and stop solution to enhance and stabilize signal. The assay is performed in standard black microplates (microplates not provided in kit) and is read on standard fluorometers.

Neuraminidase Assay Compares to NISN Protocols
The NA-Fluor assay reagent formulations are optimized to be comparable to several well-established MUNANA-based assay protocols, such as:

  • The MUNANA substrate concentration, assay buffer formulation and assay conditions are consistent with NISN IC-50 determination protocols
  • Data generated using the NA-Fluor assay corresponds to data generated with established MUNANA based protocols

These key parameters enable investigators to compare data acquired during current NI resistance surveillance screens using the NA-Fluo™ assay to data acquired using their previous MUNANA assay protocols

Detection of NI-Resistant Virus in Mixed Viral Populations
The large shift in IC-50 values between sensitive and oseltamivir-resistant virus using the NA-Fluor assay enables detection of mutant virus in mixed viral samples. This capability is critical for identifying resistant virus in clinical isolates presenting mixed populations of resistant and sensitive virus during NI susceptibility surveillance.

Flexibility for Screening Neuraminidase Activity
The NA-Fluor assay provides an easy, flexible format for the screening of several to several hundred viral isolates, or the screening of thousands of compounds during high-throughput lead discovery with quality data at high confidence levels. With superior performance, the assay has demonstrated a Z' of 0.78 to 0.8, making it strongly capable for use in high-throughput screening. The fluorescent reaction product remains stable for hours at room temperature after the assay is complete, enabling read-time flexibility and comparable data from first plate to last. The assay signal remains nearly constant and IC-50 values are identical from data collected up to 4 hours at room temperature and up to 4 days at 4°C after assay termination. The NA-Fluor assay has been optimized as an end-point assay run at 37°C for one hour following NI pre-incubation. However, the rate of MUNANA substrate turnover remains linear for more than 2 hours with viral neuraminidase, allowing the assay to be performed for as little as 20 minutes to save time or for as long as 2 hours to increase signal output. The assay can also be conducted in real-time without the addition of stop solution for those investigators who want to perform their own assay development or monitor rates of substrate turnover in the presence of inhibitor.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage This kit contains NA-Fluor™ MUNANA Substrate (4-(methylumbelliferyl)-N-acetylneuraminic acid), NA-Fluor™ 2X Assay Buffer, and NA-Fluor™ Stop Solution. Store the substrate at -20°C in the dark. Other components can be stored at 2 to 8°C. Enough reagent for ten 96-well microplate assays is supplied.
Detection Method Fluorescence
For Use With (Application) Influenza Neuraminidase Assay
For Use With (Equipment) Fluorometer
Product Type Neuraminidase Assay
Form Lyophilized
Format 96-well plate
Product Line NA-Fluor, NovaBright
Quantity 960 assays
Shipping Condition Wet Ice
Substrate NA-Fluor™
Substrate Properties Chemical Substrate
Substrate Type Neuraminidase Substrate
Target Enzyme Sialidase, Neuraminidase
Throughput High-throughput Compatible
Validated Application Enzyme Assay
Emission Wavelength Range 440/460
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.