missing translation for 'onlineSavingsMsg'
Learn More

N6-(2-Hydroxyethyl)-2'-deoxyadenosine, TRC

Product Code. 30391705
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391705 25 mg
30389854 100 mg
30389599 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391705 Supplier Toronto Research Chemicals Supplier No. TRCH94198525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137058-94-7
Molecular Formula C12H17N5O4
Molecular Weight (g/mol) 295.29
Synonym 2'-Deoxy-N-(2-hydroxyethyl)adenosine
IUPAC Name (2R,3S,5R)-5-[6-(2-hydroxyethylamino)purin-9-yl]-2-(hydroxymethyl)oxolan-3-ol
SMILES O[C@H]1C[C@H](N(C=N2)C3=C2C(NCCO)=NC=N3)O[C@@H]1CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H17N5O4/c18-2-1-13-11-10-12(15-5-14-11)17(6-16-10)9-3-7(20)8(4-19)21-9/h5-9,18-20H,1-4H2,(H,13,14,15)/t7-,8+,9+/m0/s1
Formula Weight 295.1281
Chemical Name or Material N6-(2-Hydroxyethyl)-2'-deoxyadenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.