missing translation for 'onlineSavingsMsg'
Learn More

N5-(5-Hydroxy-4,6-dimethyl-2-pyrimidinyl)-L-ornithine Hydrochloride Hydrate, TRC

Product Code. 30395713
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395713 2.5 mg 2.5mg
30401178 25 mg -
30405982 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395713 Supplier Toronto Research Chemicals Supplier No. TRCH8258052.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195143-52-3
Molecular Formula C11H18N4O3 .HCl . xH2O
Molecular Weight (g/mol) 254.29
Synonym Argpyrimidine
IUPAC Name (2S)-2-amino-5-[(5-hydroxy-4,6-dimethylpyrimidin-2-yl)amino]pentanoic acid
SMILES Cc1nc(NCCC[C@H](N)C(=O)O)nc(C)c1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H18N4O3/c1-6-9(16)7(2)15-11(14-6)13-5-3-4-8(12)10(17)18/h8,16H,3-5,12H2,1-2H3,(H,17,18)(H,13,14,15)/t8-/m0/s1
Formula Weight 254.1379
Percent Purity >95%
Chemical Name or Material N5-(5-Hydroxy-4,6-dimethyl-2-pyrimidinyl)-L-ornithine Hydrochloride Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.