CAS RN 195143-52-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N5-(5-Hydroxy-4,6-dimethyl-2-pyrimidinyl)-L-ornithine Hydrochloride Hydrate, TRC
CAS: 195143-52-3 Molecular Formula: C11H18N4O3 .HCl . xH2O Molecular Weight (g/mol): 254.29 Synonym: Argpyrimidine IUPAC Name: (2S)-2-amino-5-[(5-hydroxy-4,6-dimethylpyrimidin-2-yl)amino]pentanoic acid SMILES: Cc1nc(NCCC[C@H](N)C(=O)O)nc(C)c1O