Learn More
Invitrogen™ N4BP1 Recombinant Rabbit Monoclonal Antibody (24GB120)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557110
 View more versions of this product
Product Code. 30276612
  • £126.00 / 20µL
Description
N4BP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N4BP1
Recombinant Monoclonal
Unconjugated
N4BP1
AI481586; C81621; Kiaa0615; N4BP1; Nedd4 binding protein 1; NEDD4-binding protein 1; RGD1305179
Rabbit
Affinity chromatography
RUO
291921, 80750
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB120
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6A037
N4BP1
A synthesized peptide derived from Mouse N4BP1 (aa 286-484).
20 μL
Primary
Mouse, Rat