missing translation for 'onlineSavingsMsg'
Learn More

N4-Methylcytidine, TRC

Product Code. 30010615
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010615 25 mg 25mg
30009189 5 mg 5mg
30006108 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010615 Supplier Toronto Research Chemicals Supplier No. TRCB42656825MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10578-79-7
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.24
Synonym 1-[3,4-Dihydroxy-5-(Hydroxymethyl)Oxolan-2-Yl]-4-(Methylamino)Pyrimidin-2-One
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-(methylamino)pyrimidin-2-one
SMILES CN=C1NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H15N3O5/c1-11-6-2-3-13(10(17)12-6)9-8(16)7(15)5(4-14)18-9/h2-3,5,7-9,14-16H,4H2,1H3,(H,11,12,17)/t5-,7-,8-,9-/m1/s1
Formula Weight 257.1012
Chemical Name or Material N4-Methylcytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.