CAS RN 10578-79-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N4-Methylcytidine, TRC
CAS: 10578-79-7 Molecular Formula: C10H15N3O5 Molecular Weight (g/mol): 257.24 Synonym: 1-[3,4-Dihydroxy-5-(Hydroxymethyl)Oxolan-2-Yl]-4-(Methylamino)Pyrimidin-2-One IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-(methylamino)pyrimidin-2-one SMILES: CN=C1NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O