missing translation for 'onlineSavingsMsg'
Learn More

N1-[2-(Diethylamino)ethyl]-4-nitro-1,2-benzenediamine, TRC

Product Code. 30382334
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382334 100 mg 100mg
30385505 10 mg -
30376002 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382334 Supplier Toronto Research Chemicals Supplier No. TRCD679520100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5099-39-8
Molecular Formula C12H20N4O2
Molecular Weight (g/mol) 252.31
Synonym 1-Nitro-3-amino-4-[(beta-diethylaminoethyl)amino]benzene, 1-[(2-Diethylaminoethyl)amino]-4-nitrobenzene-2-amine, 2-(beta-Diethylaminoethylamino)-5-nitroaniline, 2-[2-(Diethylamino)ethylamino]-5-nitroaniline
SMILES CCN(CC)CCNc1ccc(cc1N)[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H20N4O2/c1-3-15(4-2)8-7-14-12-6-5-10(16(17)18)9-11(12)13/h5-6,9,14H,3-4,7-8,13H2,1-2H3
Formula Weight 252.1586
Chemical Name or Material N1-[2-(Diethylamino)ethyl]-4-nitro-1,2-benzenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.