CAS RN 5099-39-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N1-[2-(Diethylamino)ethyl]-4-nitro-1,2-benzenediamine, TRC
CAS: 5099-39-8 Molecular Formula: C12H20N4O2 Molecular Weight (g/mol): 252.31 Synonym: 1-Nitro-3-amino-4-[(beta-diethylaminoethyl)amino]benzene,1-[(2-Diethylaminoethyl)amino]-4-nitrobenzene-2-amine,2-(beta-Diethylaminoethylamino)-5-nitroaniline,2-[2-(Diethylamino)ethylamino]-5-nitroaniline SMILES: CCN(CC)CCNc1ccc(cc1N)[N+](=O)[O-]