missing translation for 'onlineSavingsMsg'
Learn More

N,S-Dimethylisothiouronium hydriodide, 98%

Product Code. p-7029918
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377279 5 g 5g
11387279 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377279 Supplier Thermo Scientific Alfa Aesar Supplier No. H27439.06
Add to basket
This item is not returnable. View return policy

CAS: 41306-45-0 | C3H9IN2S | 232.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41306-45-0
Molecular Formula C3H9IN2S
Molecular Weight (g/mol) 232.083
MDL Number MFCD00192092
InChI Key UDUMWDDMSHXGGQ-UHFFFAOYSA-N
Synonym 1,2-dimethyl-2-thiopseudourea hydriodide, 1,2-dimethylisothiourea hydriodate, n,s-dimethyl-isothiourea hydroiodide, 1,2-dimethyl-isothiourea hydroiodide, n,s-dimethylisothiouronium hydriodide, iminomethylthiomethyl methylamine, iodide, 1,2-dimethyl-2-thiopseudoureahydriodide, n-methyl methylsulfanyl methanimidamide hydroiodide, n'-methyl methylsulfanyl methanimidamide hydroiodide
PubChem CID 12263909
IUPAC Name methyl N'-methylcarbamimidothioate;hydroiodide
SMILES CN=C(N)SC.I

Specifications

Melting Point 134°C to 136°C
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material N,S-Dimethylisothiouronium hydriodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.