missing translation for 'onlineSavingsMsg'
Learn More

n-Propylmagnesium chloride, 1M in MeTHF

Product Code. 15428047
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428047 500 mL 500mL
15418047 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428047 Supplier Thermo Scientific Alfa Aesar Supplier No. H51162.AP
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2234-82-4 | C3H7ClMg | 102.84 g/mol

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2234-82-4
Molecular Formula C3H7ClMg
Molecular Weight (g/mol) 102.84
MDL Number MFCD00000474
InChI Key AMXVKMXLDJXAPM-UHFFFAOYSA-M
IUPAC Name chloro(propyl)magnesium
SMILES CCC[Mg]Cl

Specifications

Quantity 500 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 102.85
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material n-Propylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.