accessibility menu, dialog, popup

UserName

CH3MgCl

CAS RN 2234-82-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 102.84
Molecular Formula: C3H7ClMg
InChi Key: RYEXTBOQKFUPOE-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CC[CH2-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results