missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrimethoxysilane, 97+%, Thermo Scientific Chemicals

£26.30 - £255.00

Chemical Identifiers

CAS 3069-40-7
Molecular Formula C11H26O3Si
Molecular Weight (g/mol) 234.41
MDL Number MFCD00043053
InChI Key NMEPHPOFYLLFTK-UHFFFAOYSA-N
Synonym trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane
PubChem CID 76485
IUPAC Name trimethoxy(octyl)silane
SMILES CCCCCCCC[Si](OC)(OC)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11365447
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042698.06
5 g
£26.30
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11355447
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042698.14
25 g
£85.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11375447
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042698.22
100 g
£255.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

n-Octyltrimethoxysilane is used in medicine field, electronic industry and polymer material. It is also used as surface modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3069-40-7
234.41
NMEPHPOFYLLFTK-UHFFFAOYSA-N
76485
CCCCCCCC[Si](OC)(OC)OC
C11H26O3Si
MFCD00043053
trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane
trimethoxy(octyl)silane

Specifications

75°C (0.1mmHg)
Colorless to Yellow
C11H26O3Si
102°C (216°F)
Soluble in common non-polar organic solvent.
CCCCCCCC[Si](OC)(OC)OC
234.41
234.41
1.417
0.907 g/mL
3069-40-7
Liquid
MFCD00043053
trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane
NMEPHPOFYLLFTK-UHFFFAOYSA-N
trimethoxy(octyl)silane
76485
≥97%
Moisture sensitive
n-Octyltrimethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c

H315-H319-H331

missing translation for 'einecsNumber' : 221-338-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only