missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrimethoxysilane, 97+%

Product Code. 11365447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365447 5 g 5g
11355447 25 g 25g
11375447 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365447 Supplier Thermo Scientific Alfa Aesar Supplier No. 042698.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3069-40-7 | C11H26O3Si | 234.41 g/mol

n-Octyltrimethoxysilane is used in medicine field, electronic industry and polymer material. It is also used as surface modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-40-7
Molecular Formula C11H26O3Si
Molecular Weight (g/mol) 234.41
MDL Number MFCD00043053
InChI Key NMEPHPOFYLLFTK-UHFFFAOYSA-N
Synonym trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane
PubChem CID 76485
IUPAC Name trimethoxy(octyl)silane
SMILES CCCCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 75°C (0.1mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 5 g
Flash Point 102°C (216°F)
Solubility Information Soluble in common non-polar organic solvent.
Formula Weight 234.41
Percent Purity ≥97%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.907 g/mL
Chemical Name or Material n-Octyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.