missing translation for 'onlineSavingsMsg'
Learn More

N,N-[Iminobis(trimethylene)]bis-D-gluconamide, TRC

Product Code. 30397072
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397072 5 g
30395476 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397072 Supplier Toronto Research Chemicals Supplier No. TRCI4005005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86303-20-0
Molecular Formula C18 H37 N3 O12
Molecular Weight (g/mol) 487.5
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxy-N-[3-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propylamino]propyl]hexanamide
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)NCCCNCCCNC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H37N3O12/c22-7-9(24)11(26)13(28)15(30)17(32)20-5-1-3-19-4-2-6-21-18(33)16(31)14(29)12(27)10(25)8-23/h9-16,19,22-31H,1-8H2,(H,20,32)(H,21,33)/t9-,10-,11-,12-,13+,14+,15-,16-/m1/s1
Formula Weight 487.2377
Chemical Name or Material N,N-[Iminobis(trimethylene)]bis-D-gluconamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.