CAS RN 86303-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-[Iminobis(trimethylene)]bis-D-gluconamide, TRC
CAS: 86303-20-0 Molecular Formula: C18 H37 N3 O12 Molecular Weight (g/mol): 487.5 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxy-N-[3-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propylamino]propyl]hexanamide SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)NCCCNCCCNC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO