missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylphthalamide, TRC

Product Code. 30369119
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369119 5 g
30387984 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369119 Supplier Toronto Research Chemicals Supplier No. TRCD4779705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19532-98-0
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.21
Synonym N1,N2-dimethyl-1,2-benzenedicarboxamide, N,N'-Dimethylphthalamide, N,N'-Dimethylphthalic Diamide, N,N'-Dimethyl-phthalamide
IUPAC Name N1,N2-dimethylphthalamide
SMILES O=C(NC)C=1C=CC=CC1C(=O)NC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12N2O2/c1-11-9(13)7-5-3-4-6-8(7)10(14)12-2/h3-6H,1-2H3,(H,11,13)(H,12,14)
Formula Weight 192.09
Chemical Name or Material N,N'-Dimethylphthalamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.