CAS RN 19532-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N'-Dimethylphthalamide, TRC
CAS: 19532-98-0 Molecular Formula: C10H12N2O2 Molecular Weight (g/mol): 192.21 Synonym: N1,N2-dimethyl-1,2-benzenedicarboxamide,N,N'-Dimethylphthalamide,N,N'-Dimethylphthalic Diamide,N,N'-Dimethyl-phthalamide IUPAC Name: N1,N2-dimethylphthalamide SMILES: O=C(NC)C=1C=CC=CC1C(=O)NC