missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2-phenylbutanamide, TRC

Product Code. 30381329
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381329 50 mg
30387152 250 mg
30373770 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381329 Supplier Toronto Research Chemicals Supplier No. TRCE90333050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92321-53-4
Molecular Formula C14 H21 N O
Molecular Weight (g/mol) 219.32
Synonym N,N,α-Triethylbenzeneacetamide, N,N-diethyl-2-phenylbutyramide (6CI,7CI), N,N-Diethyl-2-phenylbutanamide
IUPAC Name N,N-diethyl-2-phenylbutanamide
SMILES CCC(C(=O)N(CC)CC)c1ccccc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H21NO/c1-4-13(12-10-8-7-9-11-12)14(16)15(5-2)6-3/h7-11,13H,4-6H2,1-3H3
Formula Weight 219.1623
Chemical Name or Material N,N-Diethyl-2-phenylbutanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.