CAS RN 92321-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N,N-Diethyl-2-phenylbutanamide, TRC
CAS: 92321-53-4 Molecular Formula: C14 H21 N O Molecular Weight (g/mol): 219.32 Synonym: N,N,α-Triethylbenzeneacetamide,N,N-diethyl-2-phenylbutyramide (6CI,7CI),N,N-Diethyl-2-phenylbutanamide IUPAC Name: N,N-diethyl-2-phenylbutanamide SMILES: CCC(C(=O)N(CC)CC)c1ccccc1
2
N,N-Diethyl-2-phenylbutanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.