missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Diacetyl-L-cystine (>90%), TRC

Product Code. 17938774
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17938774 5 g 5g
17939184 1 g 1g
17938694 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938774 Supplier Toronto Research Chemicals Supplier No. TRCD3125005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5545-17-5
Molecular Formula C10 H16 N2 O6 S2
Molecular Weight (g/mol) 324.37
Synonym N,N'-Diacetyl-L-cystine, Acetylcysteine Imp. C (EP)
IUPAC Name (2R)-2-acetamido-3-[[(2R)-2-acetamido-2-carboxyethyl]disulfanyl]propanoic acid
SMILES CC(=O)N[C@@H](CSSC[C@H](NC(=O)C)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C10H16N2O6S2/c1-5(13)11-7(9(15)16)3-19-20-4-8(10(17)18)12-6(2)14/h7-8H,3-4H2,1-2H3,(H,11,13)(H,12,14)(H,15,16)(H,17,18)/t7-,8-/m0/s1
Quantity 5 g
Formula Weight 324.045
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N,N'-Diacetyl-L-cystine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.