CAS RN 5545-17-5
CAS RN 5545-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N,N'-Diacetyl-L-cystine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N,N’-Diacetyl-L-cystine (>90%), TRC
CAS: 5545-17-5 Molecular Formula: C10 H16 N2 O6 S2 Molecular Weight (g/mol): 324.37 Synonym: N,N'-Diacetyl-L-cystine,Acetylcysteine Imp. C (EP) IUPAC Name: (2R)-2-acetamido-3-[[(2R)-2-acetamido-2-carboxyethyl]disulfanyl]propanoic acid SMILES: CC(=O)N[C@@H](CSSC[C@H](NC(=O)C)C(=O)O)C(=O)O