missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-butylthiourea, 98%

Product Code. 11422808
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11422808

Brand: Thermo Scientific Alfa Aesar A16783.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-46-6 | C9H20N2S | 188.333 g/mol

N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-46-6
Molecular Formula C9H20N2S
Molecular Weight (g/mol) 188.333
MDL Number MFCD00004926
InChI Key KFFQABQEJATQAT-UHFFFAOYSA-N
Synonym 1,3-dibutyl-2-thiourea, n,n'-dibutylthiourea, thiourea, n,n'-dibutyl, pennzone b, n,n'-di-n-butylthiourea, thiate u, 1,3-di-n-butyl-2-thiourea, urea, 1,3-dibutyl-2-thio, usaf ek-2138, urea, 1,3-di-n-butyl-2-thio
PubChem CID 2723622
IUPAC Name 1,3-dibutylthiourea
SMILES CCCCNC(=S)NCCCC

Specifications

Melting Point 64°C to 67°C
Odor Odorless
Quantity 1000 g
Beilstein 507434
Solubility Information Insoluble in water.
Formula Weight 188.34
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylthiourea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.