missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(1-methylpiperid-3-yl)methylamine, 90%, Thermo Scientific™

Product Code. 10135253
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135253 250 mg Amber Glass Bottle 250mg
10752114 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135253 Supplier Thermo Scientific Maybridge Supplier No. CC66146CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 639078-61-8
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD05864531
InChI Key OUBNEVZXIXKNJE-UHFFFAOYSA-N
Synonym n-methyl-1-1-methylpiperidin-3-yl methanamine, methyl 1-methylpiperidin-3-yl methyl amine, n-methyl-1-methylpiperid-3-yl methylamine, methyl-1-methyl-piperidin-3-ylmethyl-amine, 3-methylaminomethyl-1-methylpiperidine, methyl 1-methylpiperidin-3-ylmethyl amine, methyl 1-methyl 3-piperidyl methyl amine, 3-piperidinemethanamine, n,1-dimethyl-9ci, n-methyl-1-1-methyl-3-piperidinyl methanamine
PubChem CID 16640588
IUPAC Name N-methyl-1-(1-methylpiperidin-3-yl)methanamine
SMILES CNCC1CCCN(C1)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 142.24
Percent Purity 90%
Chemical Name or Material N-methyl-(1-methylpiperid-3-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.