accessibility menu, dialog, popup

UserName

CH3HNNH3C

CAS RN 639078-61-8

IUPAC Name: methyl[(1-methylpiperidin-3-yl)methyl]amine
Synonyms: methyl[(1-methylpiperidin-3-yl)methyl]amine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: OUBNEVZXIXKNJE-UHFFFAOYNA-N
SMILES: CNCC1CCCN(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results