missing translation for 'onlineSavingsMsg'
Learn More

N-Iso Valerylglycine, TRC

Product Code. 30008790
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008790 2.5 g 2.5g
30008540 1 g 1g
30006501 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008790 Supplier Toronto Research Chemicals Supplier No. TRCI9176002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16284-60-9
Molecular Formula C7 H13 N O3
Molecular Weight (g/mol) 159.18
Synonym Glycine, N-(3-methyl-1-oxobutyl)-, Glycine, N-isovaleryl- (8CI), N-(3-Methyl-1-oxobutyl)glycine, Isovalerylglycine, N-Isovaleroylglycine, N-Isovalerylglycine, N-(3-Methylbutanoyl)glycine
IUPAC Name 2-(3-methylbutanoylamino)acetic acid
SMILES CC(C)CC(=O)NCC(=O)O

Specifications

InChI Formula InChI=1S/C7H13NO3/c1-5(2)3-6(9)8-4-7(10)11/h5H,3-4H2,1-2H3,(H,8,9)(H,10,11)
Quantity 2.5 g
Formula Weight 159.0895
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Iso Valerylglycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.