CAS RN 16284-60-9
IUPAC Name:
2-(3-methylbutanamido)acetic acid
Synonyms:
isovalerylglycine
Molecular Weight (g/mol):
159.19
Molecular Formula:
C7H13NO3
InChi Key:
ZRQXMKMBBMNNQC-UHFFFAOYSA-N
SMILES:
CC(C)CC(=O)NCC(O)=O
1
–
2
of
2
results
1
N-Iso Valerylglycine, TRC
CAS: 16284-60-9 Molecular Formula: C7 H13 N O3 Molecular Weight (g/mol): 159.18 Synonym: Glycine, N-(3-methyl-1-oxobutyl)-,Glycine, N-isovaleryl- (8CI),N-(3-Methyl-1-oxobutyl)glycine,Isovalerylglycine,N-Isovaleroylglycine,N-Isovalerylglycine,N-(3-Methylbutanoyl)glycine IUPAC Name: 2-(3-methylbutanoylamino)acetic acid SMILES: CC(C)CC(=O)NCC(=O)O
2
N-Isovaleroylglycine, MedChemExpress
MedChemExpress N-Isovaleroylglycine is an acyl glycine and could be used as a biomarker for the predispositon for weight gain and obesity.