missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-4-aminobiphenyl, TRC

Product Code. 30395679
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395679 10 mg -
30394336 5 mg -
30406800 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395679 Supplier Toronto Research Chemicals Supplier No. TRCH76750010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6810-26-0
Molecular Formula C12 H11 N O
Molecular Weight (g/mol) 185.22
Synonym [1,1'-Biphenyl]-4-amine, N-hydroxy-, Hydroxylamine, N-4-biphenylyl- (7CI,8CI), N-Hydroxy[1,1'-biphenyl]-4-amine, 4-(Hydroxylamino)biphenyl, 4-Hydroxyaminobiphenyl, 4-Phenyl-N-hydroxybenzamine, N-Biphenyl-4-ylhydroxylamine, N-Hydroxy-4-aminobiphenyl
IUPAC Name N-(4-phenylphenyl)hydroxylamine
SMILES ONc1ccc(cc1)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H11NO/c14-13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13-14H
Formula Weight 185.0841
Chemical Name or Material N-Hydroxy-4-aminobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.