CAS RN 6810-26-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Hydroxy-4-aminobiphenyl, TRC
CAS: 6810-26-0 Molecular Formula: C12 H11 N O Molecular Weight (g/mol): 185.22 Synonym: [1,1'-Biphenyl]-4-amine, N-hydroxy-,Hydroxylamine, N-4-biphenylyl- (7CI,8CI),N-Hydroxy[1,1'-biphenyl]-4-amine,4-(Hydroxylamino)biphenyl,4-Hydroxyaminobiphenyl,4-Phenyl-N-hydroxybenzamine,N-Biphenyl-4-ylhydroxylamine,N-Hydroxy-4-aminobiphenyl IUPAC Name: N-(4-phenylphenyl)hydroxylamine SMILES: ONc1ccc(cc1)c2ccccc2