missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecanal, TRC

Product Code. 30397481
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397481 1 g 1g
30394093 500 mg -
30407629 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397481 Supplier Toronto Research Chemicals Supplier No. TRCH2787501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-90-3
Molecular Formula C17H34O
Molecular Weight (g/mol) 254.45
Synonym Heptadecanal, 1-Heptadecanal, Margaric Aldehyde
IUPAC Name heptadecanal
SMILES CCCCCCCCCCCCCCCCC=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h17H,2-16H2,1H3
Formula Weight 254.261
Chemical Name or Material n-Heptadecanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.