accessibility menu, dialog, popup

UserName

H3CO

CAS RN 629-90-3

IUPAC Name: heptadecanal
Synonyms: heptadecanal
Molecular Weight (g/mol): 254.46
Molecular Formula: C17H34O
InChi Key: PIYDVAYKYBWPPY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results