missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Leurosine(>85%), TRC

Product Code. 30402990
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402990 5 mg -
30393979 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402990 Supplier Toronto Research Chemicals Supplier No. TRCF7006505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54022-49-0
Molecular Formula C46 H54 N4 O10
Molecular Weight (g/mol) 822.94
Synonym Vincaleukoblastine, 4'-deoxy-3',4'-epoxy-22-oxo-, (3'α,4'α)-, (3'α,4'α)-4'-Deoxy-3',4'-epoxy-22-oxovincaleukoblastine, 1H-Indolizino[8,1-cd]carbazole, vincaleukoblastine deriv., 2H-3,13-Methanooxireno[9,10]azacycloundecino[5,4-b]indole, vincaleukoblastine deriv., 22-Oxoleurosine, F-Leurosine, Formylleurosine, Leuroformine, N-Demethyl-N-formylleurosine, N-Formyl N-demethylleurosine, N-Formylleurosine, NSC 269419, Vinformide
SMILES CC[C@@]12CN3CCc4c([nH]c5ccccc45)[C@@](C[C@@H](C3)[C@@H]1O2)(C(=O)OC)c6cc7c(cc6OC)N(C=O)[C@H]8[C@](O)([C@H](OC(=O)C)[C@]9(CC)C=CCN%10CC[C@]78C9%10)C(=O)OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C46H54N4O10/c1-7-42-15-11-17-49-19-16-44(37(42)49)30-20-31(34(56-4)21-33(30)50(25-51)38(44)46(55,41(54)58-6)39(42)59-26(3)52)45(40(53)57-5)22-27-23-48(24-43(8-2)36(27)60-43)18-14-29-28-12-9-10-13-32(28)47-35(29)45/h9-13,15,20-21,25,27,36-39,47,55H,7-8,14,16-19,22-24H2,1-6H3/t27-,36-,37?,38+,39+,42+,43+,44+,45-,46-/m0/s1
Formula Weight 822.384
Chemical Name or Material N-Formyl Leurosine (Vincristine Impurity G)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.