CAS RN 54022-49-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Formyl Leurosine(>85%), TRC
CAS: 54022-49-0 Molecular Formula: C46 H54 N4 O10 Molecular Weight (g/mol): 822.94 Synonym: Vincaleukoblastine, 4'-deoxy-3',4'-epoxy-22-oxo-, (3'α,4'α)-,(3'α,4'α)-4'-Deoxy-3',4'-epoxy-22-oxovincaleukoblastine,1H-Indolizino[8,1-cd]carbazole, vincaleukoblastine deriv.,2H-3,13-Methanooxireno[9,10]azacycloundecino[5,4-b]indole, vincaleukoblastine deriv.,22-Oxoleurosine,F-Leurosine,Formylleurosine,Leuroformine,N-Demethyl-N-formylleurosine,N-Formyl N-demethylleurosine,N-Formylleurosine,NSC 269419,Vinformide SMILES: CC[C@@]12CN3CCc4c([nH]c5ccccc45)[C@@](C[C@@H](C3)[C@@H]1O2)(C(=O)OC)c6cc7c(cc6OC)N(C=O)[C@H]8[C@](O)([C@H](OC(=O)C)[C@]9(CC)C=CCN%10CC[C@]78C9%10)C(=O)OC